Glove liners, Puri ESD Palm Fit
Supplier: PURITECH
Certificates
About this item
These thin glove liners made of 100% soft nylon fabric with carbon and PU coating on the palm are suitable for handling electronics.
- Hand-specific for great fit
- Excellent dexterity
- Anti-static, prevent damaging static discharge
- Protect humans and products from static electricity
Ideal for assembling LSI, VLSI, semiconductors as well as computers and precision instruments.
Thickness of coating: 0,1 mm.
Specifications
- Catalog No:
- PURTPFG510ESD/XL
- PURTPFG500ESD/XXL
- PURTPFG500ESD/M
- PURTPFG500ESD/L
- PURTPFG500ESD/S
- PURTPFG500ESD/XL
- Model:
- Puri ESD Palm Fit
- Puri ESD Palm Fit
- Puri ESD Palm Fit
- Puri ESD Palm Fit
- Puri ESD Palm Fit
- Puri ESD Palm Fit
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Anti-static:
- +
- +
- +
- +
- +
- +