Upright microscopes, Ceti Magnum
These microscopes provide excellent quality images via the wide-field FN20 mm eyepieces.
- Robust, T-shaped base offers maximum stability
- 30° inclined eyepiece tube ensures a comfortable viewing position over a prolonged period
- Bright, white LED transmitted lighting with intensity control
- Full Köhler illumination
- Trinocular eyepiece tube as standard to accept digital microscope cameras
: Supplied with blue and green filter, two spare fuses, immersion oil (10 ml), soft rubber eyecups, dust cover, and instruction manual.
- Catalog No:
- MEDL2721.0000
- MEDL2722.0000
- MEDL2738.0000
- Type:
- Trinocular
- Trinocular
- Trinocular
- Observation method:
- Phase contrast
- Head:
- 30° inclined
- 30° inclined
- 30° inclined
- Eyepieces:
- WF 10×/20 mm
- WF 10×/20 mm
- WF 10×/20 mm
- Stage:
- 210×140 mm (movement 90×60 mm)
- 210×140 mm (movement 90×60 mm)
- 210×140 mm (movement 90×60 mm)
- Condenser:
- Brightfield Abbe - NA 1,25; focusable and centrable, with iris diaphragm
- Brightfield Abbe - NA 1,25; focusable and centrable, with iris diaphragm
- Phase contrast turret - NA 1,25; focusable and centrable, with iris diaphragm
- Focusing system:
- Coaxial coarse and fine knobs
- Coaxial coarse and fine knobs
- Coaxial coarse and fine knobs
- Illumination:
- LED (3 W)
- LED (3 W)
- LED (3 W)
Frequently Bought Together