Deflagrating spoon
Supplier: R L ENTERPRISES
Certificates
About this item
Mild steel. Deflagrating spoon, welded to a handle.
With cork and aluminium convex cap, 90 mm diameter to give safety closure.
Frequently Bought Together
Mild steel. Deflagrating spoon, welded to a handle.
With cork and aluminium convex cap, 90 mm diameter to give safety closure.