Order Entry
Northern Ireland
ContactUsLinkComponent
Test tube holder
Test tube holder
Catalog # 212-9505
Supplier:  R L ENTERPRISES
Test tube holder
Catalog # 212-9505
Supplier:  R L ENTERPRISES

Specifications

  • Length:
    130 mm
  • Pk:
    1
  • Spanning area:
    50 mm

Specifications

About this item

Spring steel, nickel plated. This holder accepts tubes up to 50 mm diameter.