Capillary columns, TRACE™ TR-Biodiesel
Supplier: Thermo Fisher Scientific
Certificates
About this item
These columns are designed for use in carbon neutral fuels development applications.
TR-BioDiesel (M) columns: For determination of residual methanol content in Biodiesel according to EN method 14110.
TR-BioDiesel (G) columns: For analysis of total glyceride content according to EN method 14105.
TR-BioDiesel (F) columns: For analysis of FAME content in biodiesel according to EN method 14103.
TR-BioDiesel (G) ASTM columns: For analysis of total glyceride content according to ASTM method D-6584.
Frequently Bought Together