Jerricans, narrow neck
Catalog # HARRCJERPP38
Supplier: HARRISON SMITH FRENCH FLINT
Specifications
- Thread:38 mm R4
- Description:Tamper evident screw cap, PP
- Pk:10
Specifications
About this item
HDPE, translucent.
- Good chemical resistance
- Good temperature resistance
- Screw caps to be ordered separately
Universal canister for industry, trade and laboratories.