Orders LinkContactUsLinkComponent
Büchner funnels
Büchner funnels
Büchner funnels
White glazed porcelain.
  • Very good chemical resistance
  • Very good chemical resistance
  • Easy to clean
Filtering funnels acc. to Büchner. For paper or membrane filtration discs.
Product Family Options
  • Capacity
    Ø
  • 55 ml
    62 mm
    1 * 1 items
    1 * 1 items - £23.70
    £23.70
    1 * 1 items
       £23.70
  • 70 ml
    77 mm
    1 * 1 items
    1 * 1 items - £40.00
    £40.00
    1 * 1 items
       £40.00
  • 110 ml
    116 mm
    1 * 1 items
    1 * 1 items - £90.60
    £90.60
    1 * 1 items
       £90.60