ContactUsLinkComponent
Hair hygrometer
Hair hygrometer
Catalog # AMARLF105562
Supplier:  Amarell
Hair hygrometer
Catalog # AMARLF105562
Supplier:  Amarell
Specifications
  • Ø:
    100 mm
  • Range:
    0 - 100% RH
  • Pk:
    1
Specifications
About this item
This hair hygrometer is used for precise measuring of relative humidity.
  • Round shape