Surface disinfectant, perform® sterile PAA
SCUE100081
- Description:Surface disinfectant, perform® sterile PAA, in ready to use spray bottle
- Seq:02
- Packed:500 ml × 10 bottle per case
- Pk:10
Sterile disinfectant for cleanroom surfaces, based on peracetic acid. Its available both as a ready to use solution and as a liquid concentrate.
- Rapid action and effective
- Aseptically filled and microbial filtered (0,2 μm)
- Colour and perfume free
- Bactericidal, fungicidal, sporicidal, virucidal (BVDV / Vaccinia)
: Use biocides safely. Always read the label and product information before use.