Water for dilution of biocides, Premier Klerwater-WFI and -DI Water
Supplier: ECOLAB CONTAMINATION CONTROL
Certificates
About this item
Sterile WFI and deionised water are supplied cleanroom processed, 0,2 µm filtered, double bagged and terminally sterilised by gamma irradiation. Suitable for the dilution of sterile biocide concentrates or the removal of residues.
Frequently Bought Together