ContactUsLinkComponent
Hand disinfectant soaps, Phagoderm
Hand disinfectant soaps, Phagoderm
Catalog # PHAG61123
Supplier:  PHAGOGENE
Hand disinfectant soaps, Phagoderm
Catalog # PHAG61123
Supplier:  PHAGOGENE
Specifications
  • Description:
    Disinfectant soap, Phagoderm, 500 ml bottle
  • Seq:
    01
  • Pk:
    15
Specifications
About this item
Skin friendly foaming solution for frequent hygienic hand wash.
  • Combination of chlorhexidine digluconate and quaternary ammonium
  • Bactericide and fungicide
  • Perfumed
Caution: Use biocides safely. Always read the label and product information before use.