ContactUsLinkComponent
Cleanroom wipes, TechClean® ABSORBWIPE™
Cleanroom wipes, TechClean® ABSORBWIPE™
Catalog # 115-2323
Supplier:  INTERTRONICS
undefined
Cleanroom wipes, TechClean® ABSORBWIPE™
Catalog # 115-2323
Supplier:  INTERTRONICS
Specifications
  • Packed:
    100 wipes/pack
  • L×W:
    100×100 mm
  • Pk:
    100
Specifications
About this item
This very soft hydroentangled polyester wipe is non abrasive and therefore ideal for sensitive surfaces.
  • High absorbency
  • Extremly soft
  • Low-Linting
  • Double-bagged to avoid contamination
  • No chemical extractables