ContactUsLinkComponent
Beakers, low form, stainless steel
Beakers, low form, stainless steel
Catalog # COLDBEA4040C
Supplier #: BEA4040C
undefined
Beakers, low form, stainless steel
Catalog # COLDBEA4040C
Supplier #: BEA4040C
Log in for availability
Select an Option
85 mm
Select an Option
400 ml
Select an Option
85 mm
Select an Option
02
Select an Option
1
About this item
Stainless steel 304 grade.
  • Made in one piece
  • Crevice and pit free
  • Polished throughout
Ideal for corrosive materials. Robust and durable. With spout.
Frequently Bought Together