ESD occupational shoes, slip-on, Zwiesel
Catalog # SCRR11118.01.51/45
Supplier: SCHURR SCHUHVERTRIEB
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Shoe size:45
- Colour:White
- Pk:1 PAIR
Specifications
About this item
These ESD shoes are made from non perforated microfibre upper with a comfortable anti-slip PU sole. They are suitable for kitchen applications.
- DRY TECH textile lining
- Interchangeable anti-static PUR foam insole
- Good profiled sole with raised heel
Colour: White
Certifications: EN 61340 (ESD); EN ISO 20347:2004