ContactUsLinkComponent
Wipes, sterile, Klerwipe™
Wipes, sterile, Klerwipe™
Catalog # SHIE3058270
undefined
Wipes, sterile, Klerwipe™
Catalog # SHIE3058270
Specifications
  • Packed:
    10 packs × 10 wipes
  • Description:
    Klerwipe™ wipe with ultrasonically sealed edges
  • L×W:
    300×300 mm
  • Pk:
    100
Specifications
About this item
These dry surface wipes are made from 100% laundered polyester (138 g/m²) with ultrasonically sealed edges.
  • Sterilised by gamma irradiation
  • Resealable pouches with easy opening, even when wearing gloves
Packaging: Double bagged pouches