Disposable syringes, 3-piece
: SEMADENI
The piston and its seal are made of silicone rubber.
- Available with centred and eccentric tip
: Conforms with ISO 7886, 2955, 5993, 10521.
Frequently Bought Together
The piston and its seal are made of silicone rubber.