Orders LinkContactUsLinkComponent
Quantitative filter papers
Quantitative filter papers
Supplier:   SMITH SCIENTIFIC
Quantitative filter papers
Supplier:  SMITH SCIENTIFIC
Certificates
Quantitative filter papers
2 Options Available Below
Product Details & Documents
Professional filter paper grade 1 for routine laboratory applications and professional filter paper grade 4 for rapid filtration with excellent retention of larger particles.
Product Family Options
Product Information
  • Grade
    Size
    Availability
    Price
  • 1
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £60.00
    £60.00
    1 * 1 items
       £60.00
  • 4
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £62.80
    £62.80
    1 * 1 items
       £62.80