ContactUsLinkComponent
Detergent, Lyptosol
Detergent, Lyptosol
Catalog # 141-0915
Supplier:  TEEPOL
undefined
Detergent, Lyptosol
Catalog # 141-0915
Supplier:  TEEPOL
Specifications
  • Description:
    Lyptosol detergent and washing up liquid
  • Packed:
    5 litre containers
  • Pk:
    2
Specifications
About this item
Liquid detergent for washing up and general cleaning.
  • Typical usage:
  • Dish washing: 1 part to 600 parts of hot water
  • General cleaning: 1 part to 250 parts of hot water
  • Vehicles: 1 part to 450 parts of warm water