Order Entry
Northern Ireland
ContactUsLinkComponent
Tape dispenser
Tape dispenser
Catalog # TIME3161
undefined
Tape dispenser
Catalog # TIME3161

Specifications

  • For Use With:
    Tapes with 25 mm width
  • Colour:
    White
  • Description:
    Weighed tape dispenser with adjustable core
  • Pk:
    1

Specifications

About this item

This dispenser is suitable for a wide variety of labeling applications. It is designed to keep the work station and drawer clutter-free.

  • Made of sturdy plastic
  • Heavy duty with weighted base
  • Adjustable for rolls with 25 mm (1”) or 76 mm (3”) cores and 12,7 mm (¹/₂”), 19 mm (³/₄”) and 25 mm (1”) widths