Anion exchange HPLC columns, ProPac™ PA1
Supplier: Thermo Fisher Scientific
Certificates
About this item
Columns are designed for the separation and purification of hydrophilic anionic proteins and peptides.
- Pellicular packing ensure high efficiency and fast mass transport
- Ideal for high resolution separations
Specifications
- Catalog No:
- 555-7261
- 555-7262
- 555-7260
- Max. pressure:
- 345 bar (5000 psi)
- 345 bar (5000 psi)
- 345 bar (5000 psi)