Orders LinkContactUsLinkComponent
Multiple adapters
Multiple adapters
Supplier:   SMITH SCIENTIFIC
Multiple adapters
Supplier:  SMITH SCIENTIFIC
Certificates
Multiple adapters
2 Options Available Below
Product Details & Documents
Borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapter, multiple neck, one parallel neck, one at 45°.
Product Family Options
Product Information
  • Cone size
    Socket size
    Availability
    Price
  • 19/26 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £130.00
    £130.00
    1 * 1 items
       £130.00
  • 24/29 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £130.00
    £130.00
    1 * 1 items
       £130.00