SilTite™ µ-Union GC Capillary Column Connectors
Supplier: Thermo Fisher Scientific
Certificates
About this item
The SilTite µ-Union is a connector for GC capillary columns, giving zero dead volume. It has low thermal mass, is only 9 mm in length and has a mass <0,5 g.
- Zero dead volume giving optimised peak shapes
- FingerTight technology for leak free installation
- Highly inert and robust
Delivery: Each SilTite µ-Union kit contains: 5× ferrules, 2× union fittings, installation jigs, installation instructions.
Frequently Bought Together