Flasks, pear shaped with standard ground joints
Supplier: SMITH SCIENTIFIC
Certificates
About this item
Borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
The conical geometry makes them ideal for small-scale reactions.
Frequently Bought Together