Flasks, pear shaped with standard ground joint
Catalog # SMIT8SFP25/B14
Supplier: SMITH SCIENTIFIC
Specifications
- Socket size:14/23 NS
- Capacity:25 ml
- Seq:01
- Pk:1
Specifications
About this item
Borosilicate glass 3,3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
The conical geometry makes them ideal for small-scale reactions.