Beakers, tall form, borosilicate glass
Catalog # SMIT153/1000
Supplier: SMITH SCIENTIFIC
Supplier #: 153/1000
Log in for availability
About this item
Borosilicate glass, with spout.
- Excellent chemical resistance
- High temperature resistance
- Autoclavable
Ideal for general laboratory use. With spout and white graduations.
Certifications: ISO 3819, DIN 12 331, CSN 70 4031
Frequently Bought Together