Specific gravity hydrometers for medium surface tension
Supplier: S BRANNAN AND SONS
Certificates
About this item
These specific gravity hydrometers compare the density of a given liquid with that of water.
Medium surface tension is between 35 and 55 mN/m.
Certifications: ISO 650 and (now superseded) 1960 issue of BS 718.
Frequently Bought Together