Order Entry
Northern Ireland
ContactUsLinkComponent
Semi-micro cuvette
Semi-micro cuvette
Catalog # PERKB063-1065
Supplier:  PerkinElmer
Semi-micro cuvette
Catalog # PERKB063-1065
Supplier:  PerkinElmer

Specifications

  • Path length:
    10 mm
  • Type:
    Semi-micro
  • Volume:
    700 µl
  • Window material:
    Quartz (Suprasil® 300)
  • Height:
    46 mm
  • Pk:
    75 mL

Specifications

About this item

Semi-micro cuvette with PTFE stopper and black sides.