Screw caps for screw neck vials
Supplier: DELTALAB
Certificates
About this item
These screw caps are made of autoclavable PP.
- Internal silicone joint of pharmaceutical quality
Frequently Bought Together
These screw caps are made of autoclavable PP.