SPE plates, 96-well, HyperSep™ Cyano
Supplier: Thermo Fisher Scientific
Certificates
About this item
A medium polarity sorbent, Thermo Scientific™ HyperSep™ Cyano Plates are less retentive than silica or diol, and are optimized for the isolation of polar compounds from nonpolar matrices.
Typical applications include:
- Retention of polar compounds from hexane and oils
- Reversed phase extraction of moderately polar compounds
Frequently Bought Together