L-(+)-Methyl-tert-leucinate hydrochloride ≥97%
Catalog # H59563.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:181.7 g/mol
Melting Pt:183 °C
Storage Temperature:Refrigerator
CAS Number:63038-27-7
Formula:C₇H₁₅NO₂·HCl
MDL Number:MFCD00040604