

4-Fluorophenethylamine 97%
Catalog # H61791.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
CAS Number:1583-88-6
:
Formula:C₈H₁₀FN
MDL Number:MFCD00134208
:139.17 g/mol
Boiling Pt:50…52 °C
Density:1,061
Flash Pt:79 °C (174 °F)
Storage Temperature:Ambient
EINECS:216-435-6
ADR:8,II
:GHS06
:GHS05