
2-Bromophenethylamine 97%
Catalog # H60739.09
Supplier: Thermo Fisher Scientific
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
Boiling Pt:253 °C
Density:1,396
Storage Temperature:Ambient
ADR:8,II
:200.08 g/mol
UN:2735
:GHS05
CAS Number:65185-58-2
Formula:C₈H₁₀BrN
MDL Number:MFCD01529874