Order Entry
Northern Ireland
ContactUsLinkComponent
 
Citric acid 1 mol/l in aqueous solution
Catalog # BDHBBDH3397-1
Supplier:  BDH US PRODUCED
CAS Number:  
undefined
Citric acid 1 mol/l in aqueous solution
Catalog # BDHBBDH3397-1
Supplier:  BDH US PRODUCED
CAS Number:  

Specifications

  • Pk:
    12
  • Seq:
    0012

Specifications

About this item

 :
 :192.13 g/mol
EINECS:201-069-1
 :GHS05
Storage Temperature:Ambient
CAS Number:77-92-9
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669