

Tin(II) chloride dihydrate 98-103% ACS
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10025-69-1
:
Formula:SnCl₂·2H₂O
MDL Number:MFCD00149863
:225.65 g/mol
Merck Index:13,08861
Boiling Pt:623 °C (1013 hPa)
Melting Pt:246 °C
Density:2,71 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-868-0
UN:3260
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Assay:98 - 103%
- Calcium:≤0.005%
- Iron:≤0.003%
- Lead:≤0.01%
- Potassium:≤0.005%
- Sodium:≤0.01%
- Solubility in hydrochloric acid:Pass
- Sulphate (P/F):Pass
Specification Test Results
Related Information
Frequently Bought Together