Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-3-fluorotoluene
2-Bromo-3-fluorotoluene ≥98%
2-Bromo-3-fluorotoluene ≥98%
H61785.14
:
Thermo Fisher Scientific
:
2-Bromo-3-fluorotoluene ≥98%
H61785.14
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
189.03 g/mol
:
118…123 °C
:
1,503
:
76 °C (169 °F)
:
261-981-0
:
GHS07
:
Ambient
:
59907-13-0
:
C₇H₆BrF
:
MFCD08458010