
2,3,4,5-Tetrafluoroaniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5580-80-3
:
Formula:C₆H₃F₄N
MDL Number:MFCD00025153
:165.09 g/mol
Boiling Pt:78 °C
Melting Pt:27…29 °C
Flash Pt:79 °C (174 °F)
Storage Temperature:Ambient
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together