
Sodium-4-bromobenzenesulphinate dihydrate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:175278-64-5
:
Formula:C₆H₄BrNaO₂S·2H₂O
MDL Number:MFCD00210139
:279.09 g/mol (243.06 anhy)
Melting Pt:>280 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together