
4-(Trimethylsilyl)-3-butyn-2-ol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6999-19-5
:
Formula:C₇H₁₄OSi
MDL Number:MFCD00190213
:142.27 g/mol
Boiling Pt:126…129 °C
Density:0,846
Flash Pt:57 °C (135 °F)
ADR:3,III
:GHS02
Related Information
Frequently Bought Together