
5'-Bromo-2'-fluoroacetophenone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:198477-89-3
:
Formula:C₈H₆BrFO
MDL Number:MFCD11226880
:217.04 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together