
2-(4-Hydroxyphenyl)propionic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:938-96-5
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00128847
:166.18 g/mol
Melting Pt:130…132 °C
EINECS:213-352-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together