
1-(4-Chlorophenyl)cyclohexanecarboxylic acid 95%
Catalog # L05336.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:238.71 g/mol
Melting Pt:151…153 °C
EINECS:261-481-2
:GHS07
Storage Temperature:Ambient
CAS Number:58880-37-8
Formula:C₁₃H₁₅ClO₂
MDL Number:MFCD00019350