Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Cyano-2-thiopheneboronic acid
5-Cyano-2-thiopheneboronic acid ≥98%
5-Cyano-2-thiopheneboronic acid ≥98%
5-Cyano-2-thiopheneboronic acid ≥98%
L18523.03
:
Thermo Fisher Scientific
:
5-Cyano-2-thiopheneboronic acid ≥98%
L18523.03
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
305832-67-1
:
:
C₅H₄BNO₂S
:
MFCD02094029
:
152.97 g/mol
:
125…128 °C
:
GHS07
:
Ambient