1-(6-Methoxy-2-naphthyl)ethanol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77301-42-9
:
Formula:C₁₃H₁₄O₂
MDL Number:MFCD01632581
:202.25 g/mol
Melting Pt:111…114 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow to light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:111 to 114 °C
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together