
Copper(II) wolframate ≥99.5% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13587-35-4
:
Formula:CuO₄W
MDL Number:MFCD00016067
:311.38 g/mol
Merck Index:14,2657
EINECS:237-022-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together