

1-(3-(Trifluoromethyl)phenoxy)-3-butyn-2-ol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:88462-65-1
:
Formula:C₁₁H₉F₃O₂
MDL Number:MFCD02683085
:230.19 g/mol
Melting Pt:30…33 °C
:GHS06
:GHS07
Frequently Bought Together