
4-Phenyl-3-butyn-2-one 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1817-57-8
:
Formula:C₁₀H₈O
MDL Number:MFCD00008776
:144.17 g/mol
Boiling Pt:75…76 °C (0,8 mmHg)
Density:0,990
Flash Pt:95 °C (203 °F)
EINECS:217-327-1
:GHS07
Related Information
Frequently Bought Together