
1-(3-Chlorophenoxy)-3-butyn-2-ol ≥97%
Catalog # B25598.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:86912-84-7
:
Formula:C₁₀H₉ClO₂
MDL Number:MFCD09909563
:196.63 g/mol
Melting Pt:36…40 °C
:GHS07
Storage Temperature:Ambient