

1-(2-Methoxyphenyl)piperazine hydrochloride ≥97%
Catalog # B21205.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:228.72 g/mol
Melting Pt:213…216 °C
Storage Temperature:Ambient
EINECS:226-762-6
ADR:6.1,III
:GHS06
:GHS07
CAS Number:5464-78-8
Formula:C₁₁H₁₇ClN₂O
MDL Number:MFCD00012762