(S)-1-Boc-2-methylpiperazine
Catalog # APOSOR59442-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:169447-70-5
:
Formula:C₁₀H₂₀N₂O₂
MDL Number:MFCD01862121
:200.2780 g/mol