
4,4-Difluoropiperidine hydrochloride ≥98%
Catalog # L19555.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:144230-52-4
:
Formula:C₅H₁₀ClF₂N
MDL Number:MFCD03095381
:157.59 g/mol
Melting Pt:∼ 172 °C
:GHS07
Storage Temperature:Ambient